Diisopropylamine ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Isopropyl-2-propanamine

M-Clarity™ quality level = MQ200

8.03646.0100 8.03646.1000 8.03646.2500 8.03646.9020
CA8.03646.0100CS 1253 CAD
CA8.03646.0100 CA8.03646.1000 CA8.03646.2500 CA8.03646.9020
Diisopropylamine ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Diisopropylamine
Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 84
Melting Pt: -61
Density: 0.72 g/cm³ (20 °C)
Flash Pt: –13.45 °C (DIN 51755 Part 1)
Storage Temperature: Ambient
MDL Number: MFCD00008862
CAS Number: 108-18-9
EINECS: 203-558-5
UN: 1158
ADR: 3 (8) II
Merck Index: 13,03219

Order Now

Specification Test Results

Assay (GC area %) ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.715 - 0.716
Water (K. F.) ≤ 0.30 %
Identity (IR) passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR