1,2-Dimethylimidazole for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,2-Dimethyl-1H-imidazole

M-Clarity™ quality level = MQ200

8.03245.0100 8.03245.0500
CA8.03245.0100CS 1800 CAD
CA8.03245.0100 CA8.03245.0500
1,2-Dimethylimidazole for synthesis, Sigma-Aldrich®
1,2-Dimethylimidazole
Formula: C₅H₈N₂
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: 38 °C
Density: 1.084 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005294
CAS Number: 1739-84-0
EINECS: 217-101-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 30 - 40 °C
Molar Mass 96.13 g/mol
Boiling Point 205 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 93 °C
Density 1.08 g/cm3 (20 °C)
pH 12.6 (500 g/l, H2O, 20 °C)
Ignition Point 480 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR