1,1-Dichloroethylene stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,1-Dichloroethene, Vinylidene chloride

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.03209.0100 8.03209.1000
CA8.03209.0100CS 1431 CAD
CA8.03209.0100 CA8.03209.1000
1,1-Dichloroethylene stabilised for synthesis, Sigma-Aldrich®
1,1-Dichloroethylene
Formula: C₂H₂Cl₂
MW: 96.94 g/mol
Boiling Pt: 31.7
Melting Pt: -122
Density: 1.21 g/cm³ (20 °C)
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011653
CAS Number: 75-35-4
EINECS: 200-864-0
UN: 1303
ADR: 3 I
Merck Index: 12,10134

Order Now

Specification Test Results

Solubility 2.5 g/l (25 °C)
Melting Point -122 °C
Molar Mass 96.94 g/mol
Boiling Point 31.2 - 31.7 °C (1013 hPa)
Vapor Pressure 665 hPa (20 °C)
Flash Point -25 °C
Refractive Index 1.4254 (20 °C, 589 nm)
Explosion Limit 8.4 - 16.5 %(V)
Density 1.21 g/cm3 (20 °C)
Ignition Point 520 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR