Diethylamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.03010.0100
8.03010.0500
8.03010.1000
8.03010.2500
8.03010.9050
CA8.03010.0100CS
1459
CAD
CA8.03010.0100
CA8.03010.0500
CA8.03010.1000
CA8.03010.2500
CA8.03010.9050
Diethylamine for synthesis, Sigma-Aldrich®
Diethylamine
Formula:
(C₂H₅)₂NH Boiling Pt: 55.5 °C (1013 hPa) Melting Pt: –50 °C Density: 0.7048 g/cm³ (20 °C) Flash Pt: –39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009032 CAS Number: 109-89-7 EINECS: 203-716-3 UN: 1154 ADR: 3 (8),II Merck Index: 13,03138 |
|
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -50 °C |
Molar Mass | 73.14 g/mol |
Boiling Point | 56 °C |
Vapor Pressure | 250 hPa (20 °C) |
Flash Point | -25 °C |
Refractive Index | 1.3864 (20 °C) |
Explosion Limit | 2.0 - 11.8 %(V) |
Density | 0.71 g/cm3 (20 °C) |
pH | 13 (100 g/l, H2O, 20 °C) |
Ignition Point | 310 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...