Urea, GR for analysis, Supelco®
Supplier: MilliporeSigma
1.08487.0500
1.08487.1000
1.08487.5000
1.08487.9025
CA1.08487.0500CS
2257
CAD
CA1.08487.0500
CA1.08487.1000
CA1.08487.5000
CA1.08487.9025
Urea, GR for analysis, Supelco®
Urea
1.08487.0500 replaces and is the same as UX0065-1. Part # Change only, Same Pack Size, Same Specifications.
1.08487.1000 will be offered as an alternative to UX0065-1. Different Pack Size, Same Specifications.
1.08487.5000 will be offered as an alternative to UX0065-5. Different Pack Size, Same Specifications.
1.08487.9025 is offered as an alternative to UX0065-20. Different Pack Size, Same Specifications.
Formula:
NH₂CONH₂ Melting Pt: 132…135 °C (lit.) Density: 1.335 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 |
|
Specification Test Results
Solubility | 590 g/l (20 °C) |
Melting Point | 133 °C |
Molar Mass | 60.06 g/mol |
Bulk Density | 720 - 760 kg/m3 |
Vapor Pressure |
|
Density | 1.34 g/cm3 (20 °C) |
pH | 9 (100 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...