Toluene, SupraSolv® for GC-ECD/FID, Supelco®

Supplier: MilliporeSigma
Danger

1.08389.1000 1.08389.2500 1.08389.4000
CA1.08389.1000CS 1588 CAD
CA1.08389.1000 CA1.08389.2500 CA1.08389.4000
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Toluene

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: C₆H₅CH₃
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.52 g/l (20 °C)
Melting Point -95 °C
Molar Mass 92.14 g/mol
Boiling Point 110.6 °C (1013 hPa)
Vapor Pressure 29 hPa (20 °C)
Flash Point 4 °C
Refractive Index 1.4968 (20 °C)
Explosion Limit 1.2 - 8 %(V)
Density 0.87 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 535 °C
Viscosity kinematic 0.7 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR