Toluene, Uvasol® for spectroscopy, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Phenylmethane
, Methylbenzene
Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.
Formula:
C₆H₅CH₃ MW: 92.14 g/mol Boiling Pt: 110.6 °C (1013 hPa) Melting Pt: –95 °C Density: 0.867 g/cm³ (20 °C) Flash Pt: 4 °C |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 UN: 1294 ADR: 3,II Merck Index: 12,09667 |
Specification Test Results
Solubility | 0.52 g/l (20 °C) |
Melting Point | -95 °C |
Molar Mass | 92.14 g/mol |
Boiling Point | 110.6 °C (1013 hPa) |
Vapor Pressure | 29 hPa (20 °C) |
Flash Point | 4 °C |
Refractive Index | 1.4968 (20 °C) |
Explosion Limit | 1.2 - 8 %(V) |
Density | 0.87 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 535 °C |
Viscosity kinematic | 0.7 mm2/s (20 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...