Thioacetamide Reag. Ph. Eur., ACS, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ethanethioamide

M-Clarity™ quality level = MQ100

1.08170.0050 1.08170.0250
CA1.08170.0050CS 19000 CAD
CA1.08170.0050 CA1.08170.0250
Thioacetamide Reag. Ph. Eur., ACS, GR for analysis, Supelco®
Thioacetamide
Formula: CH₃CSNH₂
Melting Pt: 112…114 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (argentometric) ≥ 99.0 %
Identity (IR-spectrum) passes test
Melting point 111 - 114 °C
Heavy metals (as Pb) ≤ 0.001 %
Fe (Iron) ≤ 0.0005 %
Sulfated ash ≤ 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR