N,N-Diethyl-p-phenylenediammonium sulfate, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.03121.0100 1.03121.9025
CA1.03121.0100CS 5422 CAD
CA1.03121.0100 CA1.03121.9025
N,N-Diethyl-p-phenylenediammonium sulfate, GR for analysis, Supelco®
N,N-Diethyl-p-phenylenediammonium sulphate
Formula: C₁₀H₁₈N₂O₄S
Boiling Pt: 274…275 °C (1013 hPa)
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00012993
CAS Number: 6283-63-2
EINECS: 228-500-6

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 184 - 186 °C
Molar Mass 262.33 g/mol
Bulk Density 580 kg/m3
Boiling Point 274 - 275 °C (1013 hPa)
pH 2.1 - 2.4 (10 g/l, H2O, 20 °C)
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR