1,5-Diphenylcarbazide, GR for analysis, redox indicator, Supelco®

Supplier: MilliporeSigma
Warning

1.03091.0025 1.03091.0100
CA1.03091.0025CS 6385 CAD
CA1.03091.0025 CA1.03091.0100
1,5-Diphenylcarbazide, GR for analysis, redox indicator, Supelco®
1,5-Diphenylcarbazide
Formula: C₁₃H₁₄N₄O
Boiling Pt: 454 °C (1013 hPa)
Melting Pt: 170…172 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 173 - 176 °C
Molar Mass 242.28 g/mol
Bulk Density 420 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR