2,6-Dichlorophenolindophenol sodium salt dihydrate ≥98% (dry basis), GR, GR for the determination of ascorbic acid, Supelco®

Supplier: MilliporeSigma

1.03028.0005 1.03028.0025
CA1.03028.0005CS 52193 CAD
CA1.03028.0005 CA1.03028.0025
2,6-Dichlorophenolindophenol sodium salt dihydrate ≥98% (dry basis), GR, GR for the determination of ascorbic acid, Supelco®
2,6-Dichlorophenolindophenol sodium salt hydrate

For analysis for the determination of ascorbic acid

Formula: C₁₂H₆Cl₂NNaO₂·2H₂O
Melting Pt: ? 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150015
CAS Number: 620-45-1
EINECS: 210-640-4

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Molar Mass 326.11 g/mol
Bulk Density 320 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR