Cyclohexane, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

1.02822.0500 1.02822.2500
CA1.02822.0500CS 2236 CAD
CA1.02822.0500 CA1.02822.2500
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Cyclohexane

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: C₆H₁₂
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 55 mg/l (20 °C)
Melting Point 6 °C
Molar Mass 84.16 g/mol
Boiling Point 81 °C (1013 hPa)
Vapor Pressure 103 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4264 (20 °C)
Explosion Limit 1.2 - 8.3 %(V)
Density 0.78 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 260 °C
Viscosity kinematic 1.26 mm2/s (20 °C)
Evaporation number 3.5
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR