Chlorotrimethylsilane for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: TMCS, TMSC, Trimethylchlorosilane

1.02333.0100 1.02333.0250
CA1.02333.0100CS 7018 CAD
CA1.02333.0100 CA1.02333.0250
Chlorotrimethylsilane for gas chromatography, Supelco®
Chlorotrimethylsilane
Formula: C₃H₉ClSi
MW: 108.64 g/mol
Boiling Pt: 59
Melting Pt: -58
Density: 0.8576 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000502
CAS Number: 75-77-4
EINECS: 200-900-5
UN: 1298
ADR: 3 (8) II
REACH: 01-2119457596-25

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Melting Point -58 °C
Molar Mass 108.64 g/mol
Boiling Point 57 °C (1013 hPa)
Vapor Pressure 250 hPa (20 °C)
Flash Point -18 °C
Explosion Limit 1.5 - 46 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 420 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR