Dimethyl sulfoxide, SupraSolv® for headspace gas chromatography, Supelco®

Supplier: MilliporeSigma

Synonyms: Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane

1.01900.1000 1.01900.2500 1.01900.0500
CA1.01900.1000CS 10152 CAD
CA1.01900.1000 CA1.01900.2500 CA1.01900.0500
Dimethyl sulfoxide, SupraSolv® for headspace gas chromatography, Supelco®
Dimethyl sulphoxide

SupraSolv® solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur, and USP - SupraSolv® headspace solvents are tested for all residual solvents defined by the ICH.

Formula: (CH₃)₂SO
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Molar Mass 18.4 °C
Bulk Density 78.1 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.48 (20 °C)
Explosion Limit 1.8 - 63.0 %(V)
Density 1.10 g/cm3 (20 °C)
Ignition Point 301 °C
Saturation concentration 8.0 g/m3 (20 °C)
Viscosity kinematic 2.14 mm2/s


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR