Silver sulfate, Extra Pure, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Disilver sulphateSilver(I) sulfateDisilver sulfate

1.01534.0250 1.01534.0250 1.01534.0050 1.01534.0050
CA1.01534.0050CS 154668.21 CAD
CA1.01534.0050 EM1.01534.0250 EM1.01534.0050 CA1.01534.0250
Silver sulfate, Extra Pure, Supelco®
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
MDL Number: MFCD00003407
CAS Number: 10294-26-5
UN: 3077
ADR: 9,III
Merck Index: 13,08603
https://ca.vwr.com/stibo/web/std.lang.all/94/57/2299457.jpg

Order Now

Specification Test Results

Solubility 8 g/l (25 °C)
Melting Point 655 °C
Molar Mass 311.8 g/mol
Bulk Density 1200 kg/m3
Density 5.45 g/cm3 (20 °C)
pH 5 - 6 (5 g/l, H2O, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR