Ammonium metavanadate, GR for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.01226.0100
1.01226.0250
1.01226.1000
CA1.01226.0100CS
33758
CAD
CA1.01226.0100
CA1.01226.0250
CA1.01226.1000
Ammonium metavanadate, GR for analysis, Supelco®
Ammonium metavanadate
Formula:
NH₄VO₃ Melting Pt: 200 °C Density: 2.33 g/cm³ (20 °C) |
MDL Number:
MFCD00011430 CAS Number: 7803-55-6 EINECS: 232-261-3 UN: 2859 ADR: 6.1,II Merck Index: 13,00572 |
Specification Test Results
Solubility | 5.1 g/l (20 °C) |
Melting Point | 200 °C (decomposition) |
Molar Mass | 116.97 g/mol |
Bulk Density | 1000 kg/m3 |
Density | 2.3 g/cm3 (20 °C) |
pH | 7 (5.1 g/l, H2O, 20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...