Ammonium heptamolybdate tetrahydrate, powder, Extra Pure for spectrophotometry, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate

1.01181.0250 1.01181.1000
CA1.01181.0250CS 74855 CAD
CA1.01181.0250 CA1.01181.1000
Ammonium heptamolybdate tetrahydrate, powder, Extra Pure for spectrophotometry, Supelco®
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235.86 g/mol
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Solubility 400 g/l (20 °C)
Melting Point 90 °C Elimination of water of crystallisation
Molar Mass 1235.86 g/mol
Bulk Density 800 kg/m3
Density 2.498 g/cm3 (20 °C)
pH 5.3 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR