2-Propanol, SupraSolv® for GC-ECD/FID, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

1.00998.1000 1.00998.2500
CA1.00998.1000CS 2068 CAD
CA1.00998.1000 CA1.00998.2500
2-Propanol, SupraSolv® for GC-ECD/FID, Supelco®
2-Propanol

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -89.5 °C
Molar Mass 60.1 g/mol
Boiling Point 82.4 °C (1013 hPa)
Vapor Pressure 43 hPa (20 °C)
Flash Point 12 °C
Refractive Index 1.378
Explosion Limit 2 - 13.4 %(V)
Density 0.786 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 425 °C
Evaporation number 11
Water absorption 1000 g/kg
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR