Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene

1.00965.0500 1.00965.2500
CA1.00965.0500CS 2934 CAD
CA1.00965.0500 CA1.00965.2500
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Tetrachloroethylene

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: C₂Cl₄
MW: 165.83 g/mol
Boiling Pt: 121.1
Melting Pt: -22
Density: 1.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
EINECS: 204-825-9
UN: 1897
ADR: 6.1 III
Merck Index: 12,09332
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db3fdee4b2132ae600586a38b8bd0b99e000b343&vid=8d7c6de9c4b051ba09d36b6bdcd78117b49c3612&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.16 g/l (20 °C)
Melting Point -22 °C
Molar Mass 165.83 g/mol
Boiling Point 121 °C (1013 hPa)
Vapor Pressure 19 hPa (20 °C)
Density 1.62 g/cm3 (20 °C)
pH (H2O) not applicable
Saturation concentration 126 g/m3 (20 °C) Air
Evaporation number 11
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR