Sulfanilic acid, GR analysis_for, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

MQ100

1.00686.0100 1.00686.0250
CA1.00686.0100CS 49031 CAD
CA1.00686.0100 CA1.00686.0250
Sulfanilic acid, GR analysis_for, Supelco®
Sulphanilic acid
Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) 99.0 - 102.0 %
Identity (IR-spectrum) passes test
Matter insoluble in sodium carboN/Ate solution ≤ 0.01 %
Chloride (Cl) ≤ 0.002 %
Nitrite (NO₂) ≤ 0.5 ppm
Sulfate (SO₄) ≤ 0.01 %
Heavy metals (as Pb) ≤ 0.001 %
Sulfated ash ≤ 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR