Dithiooxamide, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Ethanebis(thioamide), Rubeanic acid

M-Clarity™ quality level = MQ100

1.00629.0010 1.00629.0100
CA1.00629.0010CS 36930 CAD
CA1.00629.0010 CA1.00629.0100
Dithiooxamide, GR for analysis, Supelco®
Dithiooxamide
Formula: C₂H₄N₂S₂
MDL Number: MFCD00004941
CAS Number: 79-40-3
EINECS: 201-203-9

Order Now

Specification Test Results

Assay (alkalimetric) ≥ 98.0 %
Identity (IR-spectrum) conforms
Suitability for copper determiN/Ation conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR