Nitric acid 65%, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.00452.1000 1.00452.2500
CA1.00452.1000CS 1372 CAD
CA1.00452.1000 CA1.00452.2500
Nitric acid 65%, EMSURE® for analysis, Supelco®
Nitric acid
Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Ag (Silver) ≤ 0.01 ppm
Al (Aluminium) ≤ 0.05 ppm
Appearance passes test
As (Arsenic) ≤ 0.01 ppm
Assay (alkalimetric) ≥ 65.0 %
Au (Gold) ≤ 0.05 ppm
Ba (Barium) ≤ 0.01 ppm
Be (Beryllium) ≤ 0.01 ppm
Bi (Bismuth) ≤ 0.1 ppm
Ca (Calcium) ≤ 0.1 ppm
Cd (Cadmium) ≤ 0.01 ppm
Chloride (Cl) ≤ 0.2 ppm
Co (Cobalt) ≤ 0.01 ppm
Cr (Chromium) ≤ 0.02 ppm
Cu (Copper) ≤ 0.01 ppm
Density (d 20/20) 1.384 - 1.416
Fe (Iron) ≤ 0.1 ppm
Fluoride (F) ≤ 1 ppm
Ga (Gallium) ≤ 0.05 ppm
Ge (Germanium) ≤ 0.02 ppm
Heavy metals (as Pb) ≤ 0.2 ppm
Hg (Mercury) ≤ 0.005 ppm
Identity passes test
In (Indium) ≤ 0.02 ppm
K (Potassium) ≤ 0.1 ppm
Li (Lithium) ≤ 0.01 ppm
Mg (Magnesium) ≤ 0.05 ppm
Minimum shelf life see product label
Mn (Manganese) ≤ 0.01 ppm
Mo (Molybdenum) ≤ 0.01 ppm
Na (Sodium) ≤ 0.2 ppm
Ni (Nickel) ≤ 0.02 ppm
Pb (Lead) ≤ 0.01 ppm
Phosphate (PO₄) ≤ 0.2 ppm
Pt (Platinum) ≤ 0.1 ppm
Residue on ignition (as SO₄) ≤ 3 ppm
Sr (Strontium) ≤ 0.01 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Tested by the dithizone passes test
Ti (Titanium) ≤ 0.02 ppm
Tl (Thallium) ≤ 0.02 ppm
V (Vanadium) ≤ 0.01 ppm
Zn (Zinc) ≤ 0.02 ppm
Zr (Zirconium) ≤ 0.02 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR