N,N-Dimethylformamide, SupraSolv® for headspace gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

1.00202.2500 1.00202.0500
CA1.00202.1000CS 7440.55 CAD
CA1.00202.1000 CA1.00202.2500 CA1.00202.0500
N,N-Dimethylformamide, SupraSolv® for headspace gas chromatography, Supelco®
N,N-Dimethylformamide

SupraSolv® solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients and drug products according to ICH, Ph Eur and USP - SupraSolv® headspace solvents are tested for all residual solvents defined by the ICH.

Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -61 °C
Molar Mass 73.09 g/mol
Boiling Point 153 °C (1013 hPa)
Vapor Pressure 3.77 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.43
Explosion Limit 2.2 - 16 %(V)
Density 0.94 g/cm3 (20 °C)
pH 7 (200 g/l, H2O, 20 °C)
Ignition Point 410 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR