Acetonitrile, SupraSolv® GC-ECD/FID, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

1.00017.1000 1.00017.2500 1.00017.4000
CA1.00017.1000CS 2068 CAD
CA1.00017.1000 CA1.00017.2500 CA1.00017.4000
Acetonitrile, SupraSolv® GC-ECD/FID, Supelco®
Acetonitrile

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 C) soluble
Melting Point -45.7 C
Molar Mass 41.05
Boiling Point 81.6 C
Vapor Pressure 97 hPA
Flash Point 2 C
Refractive Index 1.34
Explosion Limit 3.0 - 17 %
Density 0.786 g/cm3
pH (H20) N/A
Ignition Point 524 C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR