Acetonitrile, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ethyl nitrileMethyl cyanideEthanoic acid nitrileEthanenitrileCyanomethane

1.00016.1000 1.00016.1000 1.00016.2500 1.00016.2500
CA1.00016.1000CS 3175.09 CAD
CA1.00016.1000 EM1.00016.2500 CA1.00016.2500 EM1.00016.1000
Acetonitrile, Uvasol® for spectroscopy, Supelco®
Acetonitrile

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
Merck Index: 13,00071
https://ca.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg

Order Now

Specification Test Results

Solubility (20 C) soluble
Melting Point -45.7 C
Molar Mass 41.05
Boiling Point 81.6 C
Vapor Pressure 97 hPA
Flash Point 2 C
Refractive Index 1.34
Explosion Limit 3.0 - 17 %
Density 0.786 g/cm3
pH (H20) N/A
Ignition Point 524 C
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR