Embedding Cassettes, FastLoad Taped Stacks, Closed Lid, Azer Scientific
Supplier: VWR International
10030-432 10030-434 10030-436 10030-438 10030-440 10030-442 10030-444 10030-446 10030-448 10030-450 10030-452
10030-432CS 1591.62 CAD
10030-432 10030-434 10030-436 10030-438 10030-440 10030-442 10030-444 10030-446 10030-448 10030-450 10030-452
Embedding Cassettes, FastLoad Taped Stacks, Closed Lid, Azer Scientific
Embedding Cassettes Tissue Processing Cassettes
FastLoad Tissue Taped Cassettes feature a recessed hinged lid that can be open and closed as often as necessary and always relocks securely.
  • Attached and closed lid
  • Recessed, hinged lid that securely refastens
  • Three frosted writing areas
  • High-density polymer resistant to chemical actions
  • Cassettes taped for automated systems
  • Anterior writing area at 45° angle
Cassettes are molded from a high-density polymer that is resistant to solvents
The recessed lid allows for more cassettes to be stacked in auotmated systems and in storage. It is attached andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable