BRAND® Bottle Adapters for Dispensette®, seripettor® and Titrette®, BrandTech®
Supplier: Brandtech
704375 704378 704388 704390 704395 704419 704424 704429 704398 704399 704391 704431 704322 704325 704328 704338 704343 704345 704396 704397
18900-210EA 65.96 CAD
18900-210 18900-212 18900-214 18900-216 18900-218 18900-220 18900-222 18900-224 18900-226 18900-228 89136-016 89177-146 18900-323 18900-325 18900-327 18900-329 18900-331 18900-333 18900-339 18900-341
BRAND® Bottle Adapters for Dispensette®, seripettor® and Titrette®, BrandTech®
Fittings and Connectors
These polypropylene and fluoropolymer (ETFE and PTFE) adapters are for use with BRAND® bottletop dispensers and burettes.
  • Most common 4 L / 1 gal. bottles have 38 mm threads
  • Most common 1 L / 1 qt. bottles have 33 mm threads
  • Special 35 mm thread is only for use with irrigation saline used in microbiology analyzers
Outer thread refers to the outside of the adapter
Note: Many bottles come with a "neck finish" specification; a three digit number that specifies how the thread terminates on the bottle. For most laboratory bottles this is irrelevant whenMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable