Water, deionized

Supplier: Hach

27249 27256 27242
CA11014-134EA 42 CAD
CA11014-134 CA11014-136 CA11014-138
Water, deionized
Water

Hach deionized (DI) water, which has had its minieral ions removed, is an essential ingredient in hundreds of applications.


  • Demineralized Type 2 Water
  • Specific Conductance: ≤1 μS/cm, Silica (SiO2): ≤30 μg/L
  • Chloride (Cl-): ≤20 μg/L
  • Hardness: Negative
  • Consumption of KMnO4: Negative

Formula: H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR