Luer Bottom Drip Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5837-06 5837-13 5837-180 5837-204 5837-23 5837-27 5837-48 5837-53 5837-58 5837-61 5837-65 5837-80 5837-08 5837-11 5837-19 5837-16 5837-46 5837-51 5837-56 5837-21 5837-63
89052-518EA 102.04 CAD
89052-518 89052-524 89052-528 89052-532 89052-534 89052-538 89052-540 89052-544 89052-550 89052-552 89052-556 89052-558 89052-560 89052-564 89052-568 89052-570 89052-576 89052-580 89052-584 89052-628 89082-670
Luer Bottom Drip Adapter, Ace Glass Incorporated
Fittings and Connectors
This Luer Bottom Drip Adapter, with or without flow regulator valve, is used with Ace-Thred columns.
  • Available with or without flow regulator valve
  • Shallow taper at upper end
  • One FETFE O-Ring and one replaceable polyethylene packing support included
The shallow taper at the upper end minimizes mixing below the packing support. Supplied with one FETFE O-Ring and one replaceable polyethylene packing support.