Ace-Thred UNF Easy Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5810-11 5810-25 5810-35
89052-148EA 215.18 CAD
89052-148 89052-150 89052-152
Ace-Thred UNF Easy Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
The Easy Adapter allows connection of small bore tubing to Ace-Threds without having to turn vessel or tubing when removing adapter from vessel, i.e., Solvent Reservoirs.
  • Fabricated from glass-filled PTFE
  • One or two female threads with helicoil on one end
  • Silicone rubber O-Ring on other end
  • O-Ring included
One end of the adapter has one or two female 1/4"-28 threads with helicoil, and the other end has a silicone rubber O-Ring for securing in Ace-Thred with Bushing. Supplied with O-Ring, not bushing (order separately).