Ecosense DO200A Handheld Dissolved Oxygen Meters, YSI
Supplier: YSI
EcoSense®
606071 606077 606072 606040
89423-010EA 1586.35 CAD
89423-010 89423-006 89423-012 89423-014
Ecosense DO200A Handheld Dissolved Oxygen Meters, YSI
Electrochemistry Meters Oxygen Meters
The DO200A is part of the EcoSense® family of products. The DO200A dissolved oxygen instrument offers fast dissolved oxygen response times in wastewater, surface water, and aquaculture applications.
  • 1-year instrument, DO electrode and cable warranty
  • IP 67 waterproof housing
  • Screw-on cap membrane
  • Low battery indicator with 500 hour battery life
  • Automatic shutoff after 30 minutes of inactivity
  • Salinity, pressure and temperature compensation
  • 4- or 10- meter cable lengths available, BOD probe available
  • 50 data set reviewable memory
Calibrates easily in a saturated air environment and displays in % or ppm (mg/L) values, giving you the best DO data quality possible. Available with 4- or 10-meter cable lengths. Audio feedback, all keys. Calibration back up. Grey color.
The DO200A (gray case) directly replaces the DO200 (blue case).
Pressure compensation: 600 - 1100 millibar (450 - 825 mmHg)
Certifications: CE compliance. IP67 waterproof rating.
Ordering information: Instrument and cables sold seperately unless ordering as a kit. Kits include (89423-010, 89423-012, or 89423-014): instrument, cable (1-, 4-, or 10-meter), probe and carrying case. One 9V battery included. 1-year instrument and electrode warranty.