SP Bel-Art Safe-Lab™ Stirrer Bearings for Tapered Joints, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F20320-1662 F20320-1661
70680-447PK 132.15 CAD
70680-447 70680-448 70680-450
SP Bel-Art Safe-Lab™ Stirrer Bearings for Tapered Joints, Bel-Art Products, a part of SP
Stirrers Stirring Tools
Stopper-Loc™ Nut allows stirrer bearing to be locked to outer ground joint when stirring viscous materials for long periods of time.
  • PTFE construction
  • Not autoclavable
Can be locked to outer ground joint for prolonged use.
Delivery information: Supplied complete with Stopper-Loc™ Nut and two Teflon® PTFE inserts. Replacement Teflon® PTFE insert with Viton® O-Ring for 10 mm Teflon® PTFE stirrer bearings available.
Order Now
SPECIFICATIONS
Environmentally Preferable