SP Bel-Art Safe-Lab™ Joint Tubing Adapters for Tapered Joints, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F20311-1718 F20311-1717
70680-384EA 301.38 CAD
70680-384 70680-386
SP Bel-Art Safe-Lab™ Joint Tubing Adapters for Tapered Joints, Bel-Art Products, a part of SP
Adapters Adapters, Joints with Tube Connectors
Stopper has Teflon® PTFE body with Viton® O-Ring.
  • PTFE body construction
  • Polypropylene screw cap with O-ring for depth adjustment
  • Threaded extracting nut allows easy adapter removal
  • Designed for use with tapered joints
  • Not autoclavable
Polypropylene screw cap on top has an inside O-Ring for depth adjustment of glass tubing. Bottom inner joint has a threaded extracting nut for easy removal of adapter. Not autoclavable.