SP Bel-Art Pinch Valve, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SCIENCEWARE®
F30848-0005 F30848-0006 F30848-0004
89042-242CS 339.76 CAD
89042-242 89042-244 65549-653
SP Bel-Art Pinch Valve, Bel-Art Products, a part of SP
Valves Pressure-Relief Valves
Simply squeeze the soft silicone body of the valve to permit flow, and release pressure to suspend the flow.
  • Control fluid flow without drips
  • Easy to use
  • Superior chemical resistance
Valves withstand approximately 16 lbs. of pressure. Made of Teflon®, stainless steel, and polypropylene for maximum chemical resistance. Steam autoclavable at 121 °C (250 °F).