SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F30890-0000 F30891-0000 F30894-0000 F30895-0000
47750-040EA 129.32 CAD
47750-040 47750-042 47750-044 47750-046
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Stopcocks
With a removable, leak-proof plug made of chemical-resistant, non-stick Teflon® PTFE, these 2- and 3-way polypropylene plastic stopcocks have barbed tubulations that accep 6.4 to 9.5 mm (¼ to ⅜”") I.D.tubing.
  • Leakproof plug made of chemically resistant PTFE
  • Polypropylene plastic stopcock construction
  • Plugs are non-sticking
  • Autoclavable
Three-way models allow connection of any two or all three arms. Pressure rating is 240KPa (35psi). Steam autoclavable at 121°C (250°F).