SP Bel-Art Straight Tubing Connectors, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H19503-0000 H19507-0000 H19508-0000 H19509-0000 H19510-0000
62866-373PK 95.93 CAD
62866-373 62866-377 62866-378 62866-379 62866-380
SP Bel-Art Straight Tubing Connectors, Bel-Art Products, a part of SP
Fittings and Connectors
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter.
  • Tight, secure connection
  • Polypropylene plastic construction
  • Chemically resistant
  • Autoclavable
Constructed of chemical-resistant polypropylene plastic for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Autoclavable.