Reaction Flask with Flange Top and Bottom Outlet, Ace Glass Incorporated
Supplier: Ace Glass
6534-04 6534-06 6534-09 6534-50 6534-52 6534-54 6534-56 6534-60 6534-62 6534-64 6534-66
89055-424EA 2080 CAD
89055-424 89055-426 89055-428 10767-902 10767-904 10767-906 10767-908 10767-910 10767-912 10767-914 10767-916
Reaction Flask with Flange Top and Bottom Outlet, Ace Glass Incorporated
Flasks Round Bottom Flasks
This is a rugged spherical reaction flask with Duran style flat flange with O-ring groove, for use with Ace Glass quick release clamp
Flasks have capacity reference graduations, and can be configured with one of several different bottom outlets. Each flask is supplied with one CAPFE (PTFE encapsulated silicone) O-ring. Plain silicone o-rings are also available. Easy Action and Piston bottom outlets have size 35/25 O-ring ball joint connection.