CurTec Polypropylene Lids for Packo Containers, Qorpak®
Supplier: Qorpak
243028 243030 242938 242937 242943 242936 242941
76339-904CS 323.61 CAD
76339-904 76339-906 76339-908 76339-910 76339-912 76339-914 76339-916
CurTec Polypropylene Lids for Packo Containers, Qorpak®
Closures Bottle Closures
Packo Lids are available with an integrated gasket, or plain for use with inner seals also (sold separately). This High Performance Packaging line provides the highest level of product protection and added value through quality, functionality, ease of use, and design.
  • Quarter-turn screw lid
  • Water vapor tight
  • Integrated tamper evident strip
  • UN certified with Packo jars
Suggested for use with pharmaceuticals, fine chemicals, food ingredients, and hazardous waste. Airtight design prevents moisture contamination.