SP Bel-Art Super Glue, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F24006-0000
19807-000CS 554.9 CAD
19807-000
SP Bel-Art Super Glue, Bel-Art Products, a part of SP
Adhesives
A drop of fast-setting Scienceware® Super Glue provides an amazingly strong repair or assembly joint.
  • Fast-setting
  • Solvent-free
  • Colorless and transparent when set
Use on metals, rubber, wood, most plastics, or semi-porous materials. Dissimilar materials may be joined as well. Scienceware® Super Glue is an ethyl-cyanoacrylate adhesive. It is solvent-free and requires no catalyst or heat. Colorless and transparent when set. Supplied 24 per case. No air shipment.