Aluminum Oxide 90 Active, Neutral, 0.063-0.200 mm, MilliporeSigma
Supplier: Merck
1.01077.1000 1.01077.9020
EM1.01077.1000EA 318.45 CAD
EM1.01077.1000 EM1.01077.9020
Aluminum Oxide 90 Active, Neutral, 0.063-0.200 mm, MilliporeSigma
Chromatography Sorbents HPLC Sorbents
Aluminum Oxide 90 Active Neutral has 70 - 230 Mesh ASTM.