SP Bel-Art Chemical Containers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H17876-0000 H17874-0000 H17875-0000 H17871-0000
16126-446PK 98.75 CAD
16126-446 16126-450 70690-002 70690-008 74950-002 10033-652
SP Bel-Art Chemical Containers, Bel-Art Products, a part of SP
Sample Containers General Purpose Containers
Containers are great for storing small quantities of liquids or solids.
  • Heavy-walled design
  • Chemically resistant
  • Easy to clean
  • Leakproof inner seal
These heavy-walled, injection molded polyethylene jars have even been used to transport dust samples from the moon. Easy-to-clean with a screw cap design featuring a leak-proof inner seal.