WaterPro BT® Water Purification Systems, Labconco®
Supplier: Labconco
9015020 9015030 9015040 9015060 9015070 9015080 9015220 9015230 9015240 9015260 9015270 9015280 9015120 9015130 9015140 9015160 9015170 9015180 9015320 9015330 9015340 9015360 9015370 9015380
89521-834EA 10815.1 CAD
89521-834 89521-836 89521-838 89521-840 89521-842 89521-844 89521-846 89521-848 89521-850 89521-852 89521-854 89521-856 89521-858 89521-860 89521-862 89521-864 89521-866 89521-868 89521-870 89521-872 89521-874 89521-876 89521-878 89521-880
WaterPro BT® Water Purification Systems, Labconco®
Water Purification Systems Pure and Ultra Pure Water Systems
The WaterPro® BT Water Purification System produces both RO and Type I water directly from tap water
The WaterPro BT delivers ultrapure Type I, up to 18.2 megohm-cm water at a typical rate of >0.5 liters/minute. Type I water is ideal for many applications including instrumental trace element analysis and mixing standardized acid/base solutions. Reverse osmosis (RO)-purified Type III water is also produced and is stored in the integral six liter tank. An outlet valve, accessible from the back, provides access to the RO water that is produced at a rate of up to 3 liters/hour. Type III water is ideal for non-cricial laboratory applications such as reagent preparation and glassware rinsing.
The benchtop design is compact and lightweight so it can easily be transported from lab to lab. To install, the WaterPro BT requires only simple connection to a tap water supply, drain and electrical source. The Filter Pack (required, sold separately) bundles all water filter technologies into one convenient twin cartridge package so installation is simple. Models with UV light, remote dispenser or both are also available.
Ordering information: Accessories are available for the WaterPro BT. FilterPack 89521-882, comprised of twin cartridges with pretreatment, reverse osmosis, activated carbon and deionization filtration, is required for operation and is sold separately. Vent Filter (1 µm) 89521-884 helps prevent bacterial contamination of the RO storage tank and is recommended but not required. Hollow Fiber Final Filter 89521-886 for attachment to the dispensing valve is also recommended but not required. It provides a barrier against particulates and bacteria. Ultrafiltration Cartridge 89521-890 also recommended but not required, is designed for installation at the Type I spigot and produces pyrogen and nuclease-free ultrapure water for up to three months.