SP Bel-Art Wash Bottles, Wide Mouth, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F11627-1000 F11628-1000 F11625-1000 F11627-0250 F11628-0250 F11625-0250 F11626-0250 16650-022 16650-253 16650-275 F11626-1000 F11620-1000 F11620-0500 F11620-0250 F11620-0125
10844-026CS 238.08 CAD
10844-026 11215-300 16650-008 16650-014 16650-016 16650-018 16650-020 16650-022 16650-253 16650-275 47748-154 70690-056 70690-078 70690-082 10033-462
SP Bel-Art Wash Bottles, Wide Mouth, Bel-Art Products, a part of SP
Bottles Wash Bottles
Low-density polyethylene wash bottles have color-coded polypropylene closures for easy identification and wide mouths for easy filling.
  • Color-coded closures for quick identification
  • Wide-mouthed design
  • LDPE construction
Choose from Natural, Red, Yellow, Blue, and Green caps in 250, 500, or 1,000 ml sizes. Natural caps are also available in 125 ml size.
Order Now
SPECIFICATIONS
Environmentally Preferable