SP Bel-Art Wash Bottles, Red, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F11650-0016 F11651-0016
16649-998CS 356.02 CAD
16649-998 47748-158
SP Bel-Art Wash Bottles, Red, Bel-Art Products, a part of SP
Bottles Wash Bottles
The bright red color of these wash bottles helps identify hazardous contents.
  • LDPE bottle construction
  • Polypropylene closure included
  • Bright red color identifies hazardous contents
Bottles are manufactured from low-density polyethylene and are supplied with a polypropylene closure.