Heating Mantle, Pear Shaped, Ace Glass Incorporated
Supplier: Ace Glass
9516-02 9516-04 9516-06 9516-08 9516-10 9516-12
89075-030EA 607.67 CAD
89075-030 89075-032 89075-034 89075-036 89075-038 89075-040
Heating Mantle, Pear Shaped, Ace Glass Incorporated
Heaters Heating Mantles
Heating Mantle for use with pear-shaped flasks
Mantle withstands 400 Degrees Celsius internal operating temperatures. Supplied with 4 foot 2 wire cord with locking connector. Most mantles are less than 115 volts and must be operated through a power control. Use with the following power controllers,Ace Glasses' Solid State Voltage Controller, Powerstat Variable Transformer, or POWERSTAT. Not available in 230V.