Vacuum Manifold with Ace-Thred Connections, Ace Glass Incorporated
Supplier: Ace Glass
8734-07 8734-10 8734-14 8734-20
89063-780EA 1065.17 CAD
89063-780 89063-782 89063-784 89063-786
Vacuum Manifold with Ace-Thred Connections, Ace Glass Incorporated
Manifolds
Glass Manifold with #15 Ace-Thred at either end to allow switching vacuum connection to left or right side
Supplied with three or five 0 - 4 mm high vacuum, Easy-Action stopcocks for sample ports (additional ports can be ordered). Plugs are PTFE with nylon hooded handles and FETFE O-Rings. Ultimate vacuum of 10 - 7 can be realized with standard three o-rings. Overall length of glass manifold is: 9 - 1/8" (232 mm) for three ports; 13 - 5/8" (346 mm) for five ports. Supplied with “Ace-Safe” serrated connector for one end to connect vacuum line
Delivery information: Complete item consistsMore Product Information