Color Stability Tube, Ace Glass
Supplier: Ace Glass
8450-02 8450-04 8450-10
89063-040EA 285.14 CAD
89063-040 89063-042 89063-044
Color Stability Tube, Ace Glass
Tubes Centrifuge Tubes
Used in tentative AOCS Method L-15A-58 for measuring the processing color stability of fatty acids.