Stopcock, Three Way, High Vacuum, Ace Glass Incorporated
Supplier: Ace Glass
8196-250 8196-253 8196-56 8196-58
89062-640EA 589.67 CAD
89062-640 89062-642 89062-644 89062-646
Stopcock, Three Way, High Vacuum, Ace Glass Incorporated
Stopcocks
Three-way high vacuum stopcock for use in combining materials in proportion or as a common feed distributed in proportion
Easy action plugs act independently of each other and are connected to a common tubulation through a variable opening. Supplied with FETFE o-rings.