Microscale Conical Reaction Vials, Ace Glass Incorporated
Supplier: Ace Glass
9591-16 9591-21 9591-23 9591-46 9591-47 9591-48
89075-266EA 79.86 CAD
89075-266 89075-268 89075-270 89075-272 89075-274 89075-276
Microscale Conical Reaction Vials, Ace Glass Incorporated
Vials
Microscale, thin-wall reaction vial.
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The Ext.Ø of the top joint has an external thread to facilitate an O-ring and open-top phenolic cap. The internal or Int.Ø at the top is a ground standard taper 14/10 outer joint. This thin-walled vial has better heat transfer for the more demanding research project. Vials with side ports are supplied with holed cap and septum that facilitates syringe retrieval or injection for the port.
Delivery information: Supplied with threaded cap with hole, one PTFE faced silicone rubber septum and one O-ring. Outer joint for side port vials are also supplied with holed cap, PTFE-faced silicone rubber septum and FETFE O-ring.