Micro Reaction Vials, Conical Bottom, Ace Glass Incorporated
Supplier: Ace Glass
9590-02 9590-03 9590-04 9590-10 9590-11 9590-14 9590-15 9590-19 9590-20
89075-226EA 243.3 CAD
89075-226 89075-228 89075-230 89075-232 89075-234 89075-236 89075-238 89075-240 89075-242
Micro Reaction Vials, Conical Bottom, Ace Glass Incorporated
Vials
Microscale, heavy-wall reaction vial.
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The Ext.Ø. of the top joint has an external thread to facilitate an O-ring and open-top phenolic cap. The internal or Int.Ø. at the top is a ground outer joint. This type connection eliminates the need for clamps while offering the positive leaktight seal of ground glass joints.
Replace O-ring in cap with PTFE-faced silicone rubber septa and vial can be capped for storage.
Delivery information: Each vial supplied with threaded cap with hole, one PTFE faced silicone rubber septum and one O-ring for making 'O-Ring-CAP-SEAL.'